Pentoxifylline pharmaceutical secondary standard, certified reference material 1 * 500 mg
Catalog # PHR3218-500MG
Supplier: Merck
Pentoxifylline pharmaceutical secondary standard, certified reference material 1 * 500 mg
Catalog # PHR3218-500MG
Supplier: Merck
Frequently Bought Together
Sodium hypochlorite (2,5% Cl₂) in aqueous solution Reag. Ph. Eur. 1081600
Supplier: VWR Chemicals
pH buffer solutions, certified (DKD)
Supplier: Mettler - Toledo
pH buffer solutions
Supplier: Mettler - Toledo
Buffer solutions, according to EP and USP, Certipur®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
VWR®, Sulphate 1000 mg/L in water standard for ion chromatography
Supplier: VWR Chemicals
pH buffer, single-use, SINGLET™
Supplier: Hach
BUFFER PH 4,01 DOSING BOTTLE 1 * 500 mL
Supplier: HAMILTON BONADUZ
Pharmaceutical secondary standard, Caffeine, (CRM), Supelco®
Supplier: Merck
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
pH buffers, colour coded, reference standards, Supelco®
Supplier: Merck
Turbidity standards, Stablcal®
Supplier: Hach