Sumatriptan pharmaceutical secondary standard, certified reference material 1 * 100 mg
Catalog # PHR2579-100MG
Supplier: Merck
Sumatriptan pharmaceutical secondary standard, certified reference material 1 * 100 mg
Catalog # PHR2579-100MG
Supplier: Merck
Frequently Bought Together
PAH-Mix 9 deuterated 100 ng/µl 1 * 1 mL
Supplier: EHRENSTORFER
2,4-DICHLOROPHENOL 1 * 200 mg
Supplier: Merck
Tolfenpyrad certified 1 * 0,1 g
Supplier: EHRENSTORFER
CLOZAPINE PEAK IDENTIFICATION 1 * 0,41 mg
Supplier: EDQM
alpha-HCH D6 certified 1 * 10 mg
Supplier: EHRENSTORFER
Supelco® Organic Reference Standard, (±)-Enterolactone
Supplier: Merck
Ciclopirox Related Compounds, Reference Standard
Supplier: USP
Colour Standard Gardner 5 1 * 100 mL
Supplier: REAGECON
Olopatadine Related Compounds, Reference Standard
Supplier: USP
(±)-2-Butanol
Supplier: EHRENSTORFER
Supelco® Organic Reference Standard, Naphthalene-d₈ solution
Supplier: Merck
tert.-Butylhydroquinone certified 1 * 0,25 g
Supplier: EHRENSTORFER