IMAGEN CHLAMYDIA KIT 1 * 50 Tests
Catalog # OXOIK610111-2
Supplier: THERMO FISHER DIAGNOSTICS
Frequently Bought Together
FIX RAL 555 2.5L 1 * 2,5 L
Supplier: RAL
Elastin stain solution acc. Weigert, Sigma-Aldrich®
Supplier: Merck
Chinese ink, green, Q Path® stain for histology
Supplier: VWR Chemicals
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach
BOND EPITOPE RETRIEVAL SOLUTION 2 1 * 1 L
Supplier: LEICA MICROSYSTEMS
Bromocresol green sodium salt in ACS Reagent grade water ≥90% (by HPLC), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Crystal violet 1% in aqueous solution, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Reagents for ammonia determination using Lovibond® MD photometers
Supplier: Lovibond
Hydrochloric acid 2 mol/l (2 N)
Supplier: PROLABO CMP