Order Entry
France
ContactUsLinkComponent
Concept frame, manual height adjustment from 670 - 1100 mm rall 7035 grey, ESD, up to 500KG, 2000 x 750 mm 1 * 1 items
Catalog # SOVE10046034P
Supplier:  TRESTON
Concept frame, manual height adjustment from 670 - 1100 mm rall 7035 grey, ESD, up to 500KG, 2000 x 750 mm 1 * 1 items
Catalog # SOVE10046034P
Supplier:  TRESTON
Frequently Bought Together