CAP CRIMP 8MM SILVER ALUMINUM BLUE SILICONE / CLEAR PTFE SOFT 100/PACK 1 * 100 items
Catalog # HYPE6PCC8ST101
Supplier: Thermo Fisher Scientific
CAP CRIMP 8MM SILVER ALUMINUM BLUE SILICONE / CLEAR PTFE SOFT 100/PACK 1 * 100 items
Catalog # HYPE6PCC8ST101
Supplier: Thermo Fisher Scientific
Frequently Bought Together
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Qualitative Filter Papers
Supplier: VWR Collection
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Snap Cap Vials
Supplier: VWR Collection
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals