Order Entry
France
ContactUsLinkComponent
Syringe Filter, PTFE, 0.45 um, 30 mm 1 * 30 items
Catalog # STMC200-0585
undefined
Syringe Filter, PTFE, 0.45 um, 30 mm 1 * 30 items
Catalog # STMC200-0585
Frequently Bought Together