Order Entry
France
Orders LinkContactUsLinkComponent
 

Propionic acid ≥99.7%, GPR RECTAPUR®

200 L
Catalog # 87062.550
Corrosive
Flammable
Irritant
Supplier:   VWR Chemicals
Propionic acid ≥99.7%, GPR RECTAPUR®
Catalog # 87062.550
Supplier:  VWR Chemicals
CAS Number:  79-09-4
Propionic acid ≥99.7%, GPR RECTAPUR®
Propionic acid ≥99.7%, GPR RECTAPUR®

200 L

CAS Number:  79-09-4
Price
Inquire For Price
Log in for availability
1 * 200 L
Product Details & Documents
Synonyms:  Propanoic acid
  • CAS Number:79-09-4
  • Formula:C₃H₆O₂
  • MDL Number:MFCD00002756
  • Molecular Weight:74.08 g/mol
  • Boiling Pt:140…142 °C
  • Flash Pt:54 °C
  • Storage Temperature:Ambient
  • UN:3463
  • ADR:8,II
Specifications

Specification Test Results

  • Assay (GC) :
    min 99.0%
  • Identity (IR) :
    passes test
  • Density (d 20°C/4°C):
    0,993 – 0,994
  • Water (KF):
    max 0.30%
  • Appearance:
    clear liquid

Specifications

  • Pk:
    200 L

Specifications