IST - 2-FOLD LYSE 10 ML 10ML 2 PREOMICS 1 * 10 mL
Catalog # MSPPP.O.00104
Supplier: MARKET SOURCE PART PROCESS
IST - 2-FOLD LYSE 10 ML 10ML 2 PREOMICS 1 * 10 mL
Catalog # MSPPP.O.00104
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Individually Wrapped Serological Pipettes, Falcon®
Supplier: Corning
VWR® Blender Bags
Supplier: VWR Collection
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Label removal spray, Solvent 50
Supplier: HM INDUSTRIESERVICE