Order Entry
France
ContactUsLinkComponent
100 bp DNA Ladder, 1 mL. 100-1,000 bp DNA Ladder for use with the Fragment Analyzer systems and the DNF-910, DNF-935 kits or the ZAG DNA Analyzer system and the ZAG-135 kit. 1 * 1 mL
Catalog # AGLSFS-SLR910-0001
Supplier:  AGILENT
100 bp DNA Ladder, 1 mL. 100-1,000 bp DNA Ladder for use with the Fragment Analyzer systems and the DNF-910, DNF-935 kits or the ZAG DNA Analyzer system and the ZAG-135 kit. 1 * 1 mL
Catalog # AGLSFS-SLR910-0001
Supplier:  AGILENT
Frequently Bought Together