Order Entry
France
ContactUsLinkComponent
MDK DNA Ladder, 1 mL. 100-3,000 bp DNA Ladder for use with the Fragment Analyzer systems and the DNF-910CP kit. 1 * 1 mL
Catalog # AGLSFS-SLR480-0001
Supplier:  AGILENT
MDK DNA Ladder, 1 mL. 100-3,000 bp DNA Ladder for use with the Fragment Analyzer systems and the DNF-910CP kit. 1 * 1 mL
Catalog # AGLSFS-SLR480-0001
Supplier:  AGILENT
Frequently Bought Together