Order Entry
France
ContactUsLinkComponent
Markers, 1bp + 1500bp, 3.2mL. 1 bp lower marker 1,500 bp upper marker. For use with the Fragment Analyzer systems and the DNF-935 k it or the ZAG DNA Analyzer system and the ZAG-135 kit. 1 * 3,2 mL
Catalog # AGLSFA-MRK910F0003
Supplier:  AGILENT
Markers, 1bp + 1500bp, 3.2mL. 1 bp lower marker 1,500 bp upper marker. For use with the Fragment Analyzer systems and the DNF-935 k it or the ZAG DNA Analyzer system and the ZAG-135 kit. 1 * 3,2 mL
Catalog # AGLSFA-MRK910F0003
Supplier:  AGILENT
Frequently Bought Together