Order Entry
France
ContactUsLinkComponent
RNA Ladder, 20 µL. 200-6,000 nt RNA Ladder for use with the Fragment Analyzer systems and the DNF-471 kit 1 * 20 µG
Catalog # AGLSDNF-382-U020
Supplier:  AGILENT
RNA Ladder, 20 µL. 200-6,000 nt RNA Ladder for use with the Fragment Analyzer systems and the DNF-471 kit 1 * 20 µG
Catalog # AGLSDNF-382-U020
Supplier:  AGILENT