Order Entry
France
ContactUsLinkComponent
HS Large Fragment Diluent Marker, 2.4 mL. 1 bp lower marker 200,000 bp upper marker. For use with the Fragment Analyzer systems and the DNF-464 and DNF-493 kits. 1 * 2,4 mL
Catalog # AGLSDNF-381-0003
Supplier:  AGILENT
HS Large Fragment Diluent Marker, 2.4 mL. 1 bp lower marker 200,000 bp upper marker. For use with the Fragment Analyzer systems and the DNF-464 and DNF-493 kits. 1 * 2,4 mL
Catalog # AGLSDNF-381-0003
Supplier:  AGILENT
Frequently Bought Together