Order Entry
France
ContactUsLinkComponent
HS NGS Diluent Marker (1-6000 bp), 2.4 mL. 1 bp lower marker 6,000 bp upper marker. For use with the Fragment Analyzer systems and the DNF-474 kit. 1 * 2,4 mL
Catalog # AGLSDNF-373-0003
Supplier:  AGILENT
HS NGS Diluent Marker (1-6000 bp), 2.4 mL. 1 bp lower marker 6,000 bp upper marker. For use with the Fragment Analyzer systems and the DNF-474 kit. 1 * 2,4 mL
Catalog # AGLSDNF-373-0003
Supplier:  AGILENT
Frequently Bought Together