Order Entry
France
ContactUsLinkComponent
RNA Diluent Marker (15 nt), 4 mL. 15 nt lower marker. For use with the Fragment Analyzer systems and the DNF-471 kit. 1 * 4 mL
Catalog # AGLSDNF-369-0004
Supplier:  AGILENT
RNA Diluent Marker (15 nt), 4 mL. 15 nt lower marker. For use with the Fragment Analyzer systems and the DNF-471 kit. 1 * 4 mL
Catalog # AGLSDNF-369-0004
Supplier:  AGILENT