Order Entry
France
ContactUsLinkComponent
Extended Genomic DNA Ladder, 50uL. 75-48,500 bp DNA Ladder for use with the Fragment Analyzer systems and the DNF-467 kit 1 * 50 µG
  AGLSDNF-367-U050
 :  AGILENT
Extended Genomic DNA Ladder, 50uL. 75-48,500 bp DNA Ladder for use with the Fragment Analyzer systems and the DNF-467 kit 1 * 50 µG
  AGLSDNF-367-U050
 :  AGILENT
Frequently Bought Together