Order Entry
France
ContactUsLinkComponent
BF-P25 Blank Solution, 5mL. For wells in which sample is not present necessary for correct capillary alignment. For use with the Fe mto Pulse system and the FP-1002, FP-1003, FP-1101, and FP-1201 kits. 1 * 5 mL
Catalog # AGLSDNF-306-0005
Supplier:  AGILENT
BF-P25 Blank Solution, 5mL. For wells in which sample is not present necessary for correct capillary alignment. For use with the Fe mto Pulse system and the FP-1002, FP-1003, FP-1101, and FP-1201 kits. 1 * 5 mL
Catalog # AGLSDNF-306-0005
Supplier:  AGILENT
Frequently Bought Together