BF-1 Blank Solution, 8mL. For wells in which sample is not present necessary for correct capillary alignment. For use with the Frag ment Analyzer systems and the DNF-472 kit. 1 * 8 mL
Catalog # AGLSDNF-301-0008
Supplier: AGILENT
Frequently Bought Together