Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
KIT-MODERNA - COVID-19 VACCINATED SERUM (TIME SERIES), CRYOPRESERVED 1 * 1 KIT
Catalog # COVVACSRM1151M
Supplier:
Charles River Laboratories Cell Solutions
Expand image
KIT-MODERNA - COVID-19 VACCINATED SERUM (TIME SERIES), CRYOPRESERVED 1 * 1 KIT
Catalog # COVVACSRM1151M
Supplier:
Charles River Laboratories Cell Solutions
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Cell culture flasks, Falcon®
Supplier: Corning
VWR® Ultra-High Performance Centrifuge Tubes
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection