0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
France
ContactUsLinkComponent
Sterilization indicator tape, For dry heat (poupinel), dimensions: 19mm x 50m, 1x8rolls 1 * 8 items
DELT191051
:
DELTALAB
Sterilization indicator tape, For dry heat (poupinel), dimensions: 19mm x 50m, 1x8rolls 1 * 8 items
DELT191051
:
DELTALAB
Frequently Bought Together
Glove liners, Puri Top Fit
: PURITECH
Nitrile gloves, Kimtech™ Purple Nitrile™ Xtra™
: KIMBERLY CLARK
VWR®, Nitrile Gloves
: VWR Collection
VWR® Labels, Hazardous Substance, GHS Labels
: VWR Collection
Laboratory coats, C851
: Portwest
General purpose gloves, HyFlex® 11-840
: Ansell
VWR®, Laboratory Markers
: VWR Collection
Cleanroom gloves, nitrile, BioClean Nerva™
: Nitritex
Colour Lab Tapes
: GA
ESD safety shoes, slip-on, JALAS® 3520
: EJENDALS
Quantus®, Underwear Sweatshirts
: VWR Collection
Industrial ESD/touchscreen gloves, HyFlex® 11-819
: Ansell
VWR®, Cleanroom Overalls
: VWR Collection
Labels, hazardous substance, GHS pictograms
: Brady