2-PHENYL-1H-IMIDAZOLE 1 * 500 g
Catalog # APOSOR2902-500G
Supplier: Apollo Scientific
Frequently Bought Together
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
Toluene ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethylene glycol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Diethyl ether stabilised, TECHNICAL, rectified
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
Diethyl ether EMPROVE ESSENTIAL Ph Eur,BP 1 * 190 L
Supplier: MERCK PRODUCTION CHEMICALS
n-Hexane, PESTINORM® Supra Trace for MOSH/MOAH analysis
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
1-Butanol ≥98.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Diethyl ether, anhydrous ≥99% stabilised ACS
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide 99.8+% for HPLC
Supplier: Thermo Fisher Scientific