FIDAXOMICIN 1 * 100 mg
Catalog # APOSBIW4203-100MG
Supplier: Apollo Scientific
Frequently Bought Together
VWR® Ultra-High Performance Centrifuge Tubes
Supplier: VWR Collection
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Pasteur pipettes
Supplier: KARTELL
VWR®, Spreaders, L-Shaped, Standard Line
Supplier: VWR Collection
Round-bottom polystyrene tubes, disposable, Falcon®
Supplier: Corning
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
1,4-Dioxane, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
96-Well cell culture plates, Falcon®
Supplier: Corning
Transport container with screw cap
Supplier: BOETTGER PAUL
Dichloromethane stabilised, EMPLURA®
Supplier: Merck
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethylenediamine ≥99.0% (by GC), GPR RECTAPUR®
Supplier: VWR Chemicals