3-ISOPROPOXY-5-(TRIFLUOROMETHOXY)PHENYLB 1 * 5 g
Catalog # APOSPC412505-5G
Supplier: Apollo Scientific
3-ISOPROPOXY-5-(TRIFLUOROMETHOXY)PHENYLB 1 * 5 g
Catalog # APOSPC412505-5G
Supplier: Apollo Scientific
Frequently Bought Together
2-Propanol, EMPROVE® EXPERT Ph. Eur., BP, USP for preparative chromatography, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Pyrene for synthesis, Sigma-Aldrich®
Supplier: Merck
Methanol, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Diisopropyl ether stabilised for analysis
Supplier: Thermo Fisher Scientific
3-(Chloromethyl)benzoyl chloride 98%
Supplier: Thermo Fisher Scientific
4-Methyl-2-pentanone 99+% for HPLC
Supplier: Thermo Fisher Scientific
1,2-Dichlorobenzene 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform 99.8% stabilised for analysis
Supplier: Thermo Fisher Scientific
Pyridine 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
4,4'-Bis(dimethylamino)diphenylmethane 98%
Supplier: Thermo Fisher Scientific
(±)-2-Butanol ≥99.5% (by GC), Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals