5-(2,6-DIFLUOROPHENYL)ISOXAZOLE 1 * 5 g
Catalog # APOSPC9967-5G
Supplier: Apollo Scientific
Frequently Bought Together
1-Butanol ≥98.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
N,N-Dimethylformamide ≥99.9%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
n-Hexane ≥98%, SupraSolv® for GC-MS
Supplier: Merck
2,2,4-Trimethylpentane ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
1,3-Dimethyl-2-imidazolidinone ≥99.5%, PESTINORM® for headspace gas chromatography
Supplier: VWR Chemicals
Dimethyl sulphoxide ≥99.5%, BioChemica® cell culture reagent
Supplier: PanReac AppliChem
Petroleum spirit, 60…80 °C, EMSURE® for analysis, Supelco®
Supplier: Merck
Methyl ethyl ketone ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Isoamyl alcohol, TECHNICAL
Supplier: VWR Chemicals
N,N-Dimethylformamide ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanol USP, J.T.Baker®
Supplier: Avantor
Ammonia solution 35% in water for HPLC
Supplier: Thermo Fisher Scientific