METHYL 2,3-DIAMINOBENZOATE 1 * 5 g
Catalog # APOSOR472041-5G
Supplier: Apollo Scientific
Frequently Bought Together
N-Hydroxysuccinimide for synthesis, Sigma-Aldrich®
Supplier: Merck
N,N-Dimethyl-p-toluidine for synthesis, Sigma-Aldrich®
Supplier: Merck
Ethanol absolute for histology, J.T.Baker®
Supplier: Avantor
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals
Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
o-Xylene ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimethylacetamide ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform 99.8% stabilised for analysis
Supplier: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) ≥99.7%, Biotechnology Grade, Burdick & Jackson™
Supplier: Honeywell Chemicals
Ethyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Diethyl ether, anhydrous 99+% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Ethanol absolute Ph. Eur., J.T.Baker®
Supplier: Avantor