RNASE H 1 * 1 Each
Catalog # APOSBIBL0050-250U
Supplier: Apollo Scientific
Frequently Bought Together
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
Storage plates, 96-well, MASTERBLOCK®
Supplier: Greiner Bio-One
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Transfer pipettes
Supplier: RATIOLAB
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Sampling Spoons
Supplier: VWR Collection