[EN]SOFT CARE SENSITIVE 10X0.5L W2738 1 * 10 Fl
Catalog # DIVY101108207
Supplier: Diversey
Frequently Bought Together
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dichloromethane ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
tert-Butyl methyl ether ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Supplier: Thermo Fisher Scientific
Bis(2-methoxyethyl)ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 2,5 L
Supplier: PanReac AppliChem
n-Heptane ≥99% (GC (n-heptane basis)), Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Kerosene, Reagent Grade low odor, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone ACS, ISO, Reag. Ph. Eur., Burdick & Jackson™
Supplier: Honeywell Chemicals
Carbon disulphide 99.9% for HPLC
Supplier: Thermo Fisher Scientific
Tetrachloroethylene 99%, extra pure
Supplier: Thermo Fisher Scientific
1-Chlorobutane ≥99%
Supplier: Thermo Fisher Scientific