Kleenex Extra Gel Hand Sanitiser - Cassette / Clear /1 Litre 1 * 6 items
Catalog # KIMB6382-001
Supplier: KIMBERLY CLARK
Kleenex Extra Gel Hand Sanitiser - Cassette / Clear /1 Litre 1 * 6 items
Catalog # KIMB6382-001
Supplier: KIMBERLY CLARK
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Dimethyl sulphoxide, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥98.5%, TECHNICAL
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Pyridine ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
Methanol ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile ≥99%, TECHNICAL
Supplier: VWR Chemicals
Dichloromethane stabilised, EMPLURA®
Supplier: Merck
Toluene, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
2,2,4-Trimethylpentane ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals