[cis-3,4,6,7,8,8a-hexahydro-1H-pyrrolo[2,1-c][1,4]oxazin-6-yl]methanol 1 * 1 g
Catalog # PHBLPBQA859-1G
Supplier: Pharmablock (USA)
[cis-3,4,6,7,8,8a-hexahydro-1H-pyrrolo[2,1-c][1,4]oxazin-6-yl]methanol 1 * 1 g
Catalog # PHBLPBQA859-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Methanol ≥99.9% (by GC), CHROMASOLV™ LC-MS Ultra tested for UHPLC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
Water with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Methanol ≥99.8%, Ultrapure for HPLC
Supplier: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
n-Hexane ≥97%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol absolute, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Tetrahydrofuran ≥99.9% stabilised, Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl sulphoxide ≥99.5%, Reagent Grade
Supplier: Honeywell Chemicals
Butanone (Methylethylketone) pure 1 * 2,5 L
Supplier: PanReac AppliChem
Carbon disulphide 99.9% for HPLC
Supplier: Thermo Fisher Scientific
Ethanol (untaxed) ~96%, Puriss. p.a. ACS, without additive prima fine spirit, F15 (not denatured), Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclohexane ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals