ETHYL4-(TRIFLUOROMETHYL)-1H-PYRAZOLE-3-CARBOXYLATE 1 * 25 g
Catalog # PHBLPBXAA1090-25G
Supplier: Pharmablock (USA)
ETHYL4-(TRIFLUOROMETHYL)-1H-PYRAZOLE-3-CARBOXYLATE 1 * 25 g
Catalog # PHBLPBXAA1090-25G
Supplier: Pharmablock (USA)
Frequently Bought Together
Cyclohexane ≥99.8%, SupraSolv® for GC-MS
Supplier: Merck
Ethylene glycol, EMPLURA®, Supelco®
Supplier: Merck
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Phenyl isocyanate, Supelco®
Supplier: Merck
1,4-Dioxane ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimethylacetamide 99.8+% for HPLC
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Diethyl ether ≥99.8% contains inhibitor, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone ACS, ISO, Reag. Ph. Eur., Burdick & Jackson™
Supplier: Honeywell Chemicals
tert-Butanol 99.5%, extra pure
Supplier: Thermo Fisher Scientific