Rouleau étiquettes : LABEL WHITE 23 (W) x 17mm (H) 5000 pcs , 050867: PP 60 WHITE EXTRA PERMANENT - roll width : 26,00 mm core diam. 45m - distance between labels : 4,09 mm 1 * 1 ROU
Catalog # LBMI050867
Supplier: LABONORD MISCELLANEOUS
Frequently Bought Together