Order Entry
France
Orders LinkContactUsLinkComponent
VeriFluor™ Far-Red Passive Reference Dye

VeriFluor™ Far-Red Passive Reference Dye

VeriFluor™ far-red passive reference dye, 400× in water, 640/660 nm, 50 µl
Catalog # 29087-50UL
Supplier:   Biotium
VeriFluor™ Far-Red Passive Reference Dye
Catalog # 29087-50UL
Supplier:  Biotium
CAS Number:  
VeriFluor™ Far-Red Passive Reference Dye

VeriFluor™ far-red passive reference dye, 400× in water, 640/660 nm, 50 µl

CAS Number:  
Price
164,00 €
Log in for availability
1 * 50 µl
Product Details & Documents

A far-red passive reference dye for qPCR that frees up the ROX channel for use with JUN™ probes, EvaRuby™ qPCR dye, and other orange dyes.

  • An alternative to ROX for qPCR normalization in the red channel
  • Drop-in replacement for Mustang Purple™ on most instruments
  • Tolerates DTT in qPCR reactions, unlike Mustang Purple™
  • More soluble than ROX for easier mixing and more accurate results
  • Stand-alone dye solution for use with our Forget-Me-Not™ Master Mixes or other mixes

VeriFluor™ Far-Red is a far-red passive reference dye that is used to normalize the fluorescent reporter signal during qPCR and eliminate well-to-well variation. It is an alternative to normalization with ROX passive reference dye and a direct replacement for Mustang Purple™.

Specifications

Specifications

  • Pk:
    50 µl
  • Description:
    VeriFluor™ far-red passive reference dye, 400× in water
  • Ex/Em:
    640/660 nm
  • Cat. No.:
    29087-50UL
  • Storage conditions:
    Store at –10...–35 °C

Specifications