(2S)-2-{[(TERT-BUTOXY)CARBONYL]AMINO}-2-(4,4-DIFLUOROCYCLOHEXYL)ACETIC ACID 1 * 400 g
Catalog # PHBLPBLJ3451-400G
Supplier: Pharmablock (USA)
(2S)-2-{[(TERT-BUTOXY)CARBONYL]AMINO}-2-(4,4-DIFLUOROCYCLOHEXYL)ACETIC ACID 1 * 400 g
Catalog # PHBLPBLJ3451-400G
Supplier: Pharmablock (USA)
Frequently Bought Together
Trifluoroacetic acid ≥99.9% for peptide synthesis
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
1-Propanol, EMPLURA®, Supelco®
Supplier: Merck
Cyclohexane ≥99.5%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Glycerine ≥97%, TECHNICAL
Supplier: VWR Chemicals
N,N-Dimethylformamide ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
Methyl ethyl ketone ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute (untaxed) ≥99.8%, Puriss. p.a. ACS, A15, without additive, Riedel-de Haën™
Supplier: Honeywell Chemicals
Sinapic acid, Supelco®
Supplier: Merck
Acetone, J.T.Baker®
Supplier: Avantor