WIPE WYPALL L CENTREFEED 38X18,3CM 500/R 1 * 6 Roll
Catalog # KIMB7406
Supplier: KIMBERLY CLARK
Frequently Bought Together
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Wipes for delicate tasks, Kimtech™ Science
Supplier: KIMBERLY CLARK
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Screw caps for screw neck vials, N 8
Supplier: MACHEREY-NAGEL
Closing cones, Combi-Stoppers
Supplier: B. Braun
VWR® Spec-Wipe® 3, Cleanroom Wipes, Non Woven
Supplier: VWR Collection
VWR®, Bottle-Top Vacuum Filtration Systems, PVDF, SFCA, Nylon
Supplier: VWR Collection
pH electrodes, SenTix®
Supplier: WTW
VWR®, Latex Gloves
Supplier: VWR Collection
VWR® VV3, Vortex Mixer
Supplier: VWR Collection
VWR®, Bench Protectors
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
BUFFER PH 7,0 +/- 0,01 SOLUTION 25 1 * 500 mL
Supplier: REAGECON
Sodium acetate trihydrate 99+%, extra pure
Supplier: Thermo Fisher Scientific