0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
France
ContactUsLinkComponent
ACETYLSALICYLIC ANHYDRIDE 1 * 10 mg
PROOTRCA187785-10
:
LGC Standards PROMOCHEM
ACETYLSALICYLIC ANHYDRIDE 1 * 10 mg
PROOTRCA187785-10
:
LGC Standards PROMOCHEM
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
: Merck
pH buffer solutions
: Mettler - Toledo
Buffer solutions in sachets
: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
: Merck
pH buffers, ready to use
: WTW
VWR®, Conductivity standard solutions
: VWR Chemicals
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
: Merck