1-{spiro[3.3]heptan-2-yl}methanamine hydrochloride 1 * 1 g
Catalog # PHBLPBZ1440-1-1G
Supplier: Pharmablock (USA)
1-{spiro[3.3]heptan-2-yl}methanamine hydrochloride 1 * 1 g
Catalog # PHBLPBZ1440-1-1G
Supplier: Pharmablock (USA)
Frequently Bought Together
Ethanol absolute, anhydrous (max. 0.003% H₂O) ≥99.8%
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
N,N-Dimethylformamide ≥98%, TECHNICAL
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Dichloromethane ≥99.8% stabilised, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals