ENDO-CLEANER 5 L KORSOLEX 1 * 5 L
Catalog # BODE980117
Supplier: BODE CHEMIE
Frequently Bought Together
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., JP, USP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
N,N-Dimethyl-p-toluidine for synthesis, Sigma-Aldrich®
Supplier: Merck
Dimethyl Sulfoxide, 99.5% for synthesis 1 * 2,5 L
Supplier: PanReac AppliChem
Methanol ACS, gradient grade for HPLC, Burdick & Jackson™
Supplier: Honeywell Chemicals
Ethanol absolute Ph. Eur., J.T.Baker®
Supplier: Avantor
Isoamyl alcohol 98%, pure
Supplier: Thermo Fisher Scientific
3-(Chloromethyl)benzoyl chloride 98%
Supplier: Thermo Fisher Scientific
Ethyl acetate ACS, ISO, Reag. Ph. Eur., Burdick & Jackson™
Supplier: Honeywell Chemicals
Pyridine ≥99%
Supplier: Thermo Fisher Scientific
Acetonitrile, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Diisopropyl ether stabilised for analysis
Supplier: Thermo Fisher Scientific
2-Methylbutane ≥99%, Reagent Grade
Supplier: Honeywell Chemicals