WIPE WYPALL WETTASK LOW LINT 12X55PCS 1 * 12 Roll
KIMB7752
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR®, Beard Cover, PP
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
Ethanol 96% vol Ph. Eur.
: VWR Chemicals
Single-use syringes, 3-piece, HENKE-JECT®
: HENKE SASS WOLF