Recombinant Guinea pig CACNA1C protein 1 * 100 µG
Catalog # ABCAAB289760-100
Supplier: Abcam
Frequently Bought Together
Screw caps for microcentrifuge tubes
Supplier: Thermo Fisher Scientific
VWR®, Cap Sealing Mats for 96 Deep Well Plates
Supplier: VWR Collection
Microcentrifuge tubes, 0,5/1,5/2,0 ml
Supplier: Brand
Chemical bottles, wide mouth, square
Supplier: RIXIUS
CAP FOR MICRO METHOD PLATE IRR BETA 1 * 300 items
Supplier: Anicrin
Storage Boxes, PP, with Lid
Supplier: KOEHLER TECHNISCHE PRODUKTEN
Cover mats for deep well plates
Supplier: Brand
HTS TranswellRegistered Mark-96, 0.4 µm Pore Polyester Membrane Inserts 1 * 480 items
Supplier: STEMCELL Technologies
Filter plates, 96-well, for protein purification, AcroPrep™ Advance
Supplier: Cytiva (Formerly Pall Lab)
Water sampling bottles, rectangular
Supplier: DELTALAB
Cryoboxes, Max-100 CryoStore™, Nunc™
Supplier: Thermo Fisher Scientific
Tube ScrewTop 1,0ml natural V-Bottom with white patch and linear barcode sterile. For UK BioBank only- custom 1 * 480 items
Supplier: THERMO MATRIX TECHNOLOGIES
Racks and lids for storage 2D barcoded tubes, Matrix™
Supplier: THERMO MATRIX TECHNOLOGIES
Microcentrifuge tubes, 1,5 ml, in resealable bags, SuperSpin®
Supplier: Labcon
Cryoboxes, Cardboard
Supplier: RATIOLAB