(2S)-2-(TERT-BUTOXYCARBONYLAMINO)-2-(3,3-DIFLUOROCYCLOBUTYL)ACETIC ACID 1 * 25 g
PHBLPBA0087-25G
(2S)-2-(TERT-BUTOXYCARBONYLAMINO)-2-(3,3-DIFLUOROCYCLOBUTYL)ACETIC ACID 1 * 25 g
PHBLPBA0087-25G
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
: VWR Chemicals
Water USP, JP, Ph. Eur., purified
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
: VWR Chemicals