[EN]CABLE POWER EUROPEAN 10A/250V 1 * 1 ST
Catalog # ABSX5027841DE
Supplier: AB SCIEX
Frequently Bought Together
Gas detection tubes for nitrous fumes
Supplier: Dräger
Tubes and systems for active sampling
Supplier: Dräger
Gas detection tubes for sulphur dioxide
Supplier: Dräger
VWR®, Accessories for VWR® SAS Super
Supplier: VWR Collection
Accessories for Dewar vessels, Agil series
Supplier: Air Liquide
Viscosity cups, Zahn
Supplier: Industrial Physics
CAMERA EP50 1 * 1 items
Supplier: OLYMPUS OPTICAL
Accessories for buchi melting point apparatus M560 and M565
Supplier: BUCHI
Colorimetrics tubes for gas measurements
Supplier: GASTEC
Accessories for ÄKTA chromatography systems
Supplier: Cytiva
Reference electrodes, Radiometer Analytical
Supplier: Hach
TUBE NITROSE GAS 50/B 1 * 10 items
Supplier: Dräger
VWR® CO 310/CO 310 M, Conductivity Meters, Handheld
Supplier: VWR Collection
Components for spectrophotometric colorimeters, PFX<i>i</i>-195/880/950/995 series, Lovibond®
Supplier: Lovibond Tintometer