4'-CHLORODIAZEPAM-13C2,15N 1 * 1 mL
Catalog # C-235-1ML
Supplier: Merck
Frequently Bought Together
Buffer solutions as per DIN 19266-01: 2000
Supplier: WTW
Sodium carbonate Certipur 1 * 80 g
Supplier: Merck
pH Standard Solutions, IUPAC, Certified
Supplier: Hach
Bendamustine Related Compound, Reference Standard
Supplier: USP
TETRASUL 1 * 50 mg
Supplier: Merck
Nitrate standards for ISE calibration, Orion™
Supplier: Thermo Orion
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond
n-Pentacosane
Supplier: EHRENSTORFER
Supelco® Organic Reference Standard, Palm oil
Supplier: Merck
PARACETAMOL IMPURITY J 1 * 15 mg
Supplier: EDQM
VWR®, Gardner colour standards
Supplier: VWR Chemicals
Reagents for chloride determination using Lovibond® MD photometers
Supplier: Lovibond
VWR® ARISTAR®, Phosphorus Standard Solutions for ICP-MS
Supplier: VWR Chemicals