Order Entry
France
ContactUsLinkComponent
Liner, Borosilicate, 5/PK 88 mm, 3 mm ID, 1 ring restriction, filled with deactivated borosilicate wool 1 * 5 Pce
  MSPPJAS-7920-124-5
Liner, Borosilicate, 5/PK 88 mm, 3 mm ID, 1 ring restriction, filled with deactivated borosilicate wool 1 * 5 Pce
  MSPPJAS-7920-124-5
Frequently Bought Together