tert-butyl N-[(3R,4R)-3-fluoropiperidin-4-yl]carbamate 1 * 9 g
Catalog # PHBLPB08067-9G
Supplier: Pharmablock (USA)
tert-butyl N-[(3R,4R)-3-fluoropiperidin-4-yl]carbamate 1 * 9 g
Catalog # PHBLPB08067-9G
Supplier: Pharmablock (USA)
Frequently Bought Together
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol eurodenatured ≥99%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
Toluene ≥99.9% (by GC), SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
Toluene, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
n-Heptane
Supplier: Merck
Acetonitrile ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Toluene, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
1,3-Propanediol ≥99.7%, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Ethanol 90% 90% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
Aniline for synthesis, Sigma-Aldrich®
Supplier: Merck
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
2-Propanol ≥99.8% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals