2-(2-Aminoethyl)-3-(4-chlorophenyl)-3-hydroxyisoindol-1-one (hydrochloride) 1 * 1 mg
Catalog # CAYM34667-1
Supplier: Cayman Chemical
2-(2-Aminoethyl)-3-(4-chlorophenyl)-3-hydroxyisoindol-1-one (hydrochloride) 1 * 1 mg
Catalog # CAYM34667-1
Supplier: Cayman Chemical
Frequently Bought Together
96-Well Polypropylene Storage Blocks, Corning®
Supplier: Corning
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Deepwell Plates 96-Well, 2000 µl
Supplier: EPPENDORF
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Chloroform ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
Supplier: VWR Collection
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR® Cryoboxes and Dividers
Supplier: VWR Collection
, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
VWR® Disposable Microcentrifuge Tubes
Supplier: VWR Collection
n-Hexane ≥99%, PESTINORM® Supra Trace for organic trace analysis
Supplier: VWR Chemicals