PHCbi Energy Efficient Medical - Pharmaceutical Refrigerator with Inverter Control Compressor and HC Refrigerants. 550 litre interior capacity. Temperature control range 2C to 14C. Fan forced air circulation for maximum temperature uniformity. Micr 1 * 1 items
Catalog # SANYMPR-S500RH
Supplier: PHC Corporation
Frequently Bought Together