Order Entry
France
Orders LinkContactUsLinkComponent
Recycling solution for protective eyewear, Zero Waste Box™

Recycling solution for protective eyewear, Zero Waste Box™

Disposal box for protective eyewear, small, 28 L, ≤7 kg, 250×250×460 mm
Catalog # 129-2443
   Waste Reducing|Sustainable Materials
Supplier:   TERRACYCLE
Recycling solution for protective eyewear, Zero Waste Box™
Catalog # 129-2443
Supplier:  TERRACYCLE
   Waste Reducing|Sustainable Materials
Recycling solution for protective eyewear, Zero Waste Box™
Recycling solution for protective eyewear, Zero Waste Box™

Disposal box for protective eyewear, small, 28 L, ≤7 kg, 250×250×460 mm

Price
98,75 €
Log in for availability
1 * 1 Pce
Product Details & Documents

A Zero Waste Box™ is a complete and convenient solution which includes the storage, shipping and recycling of a particular type of waste that isn’t currently recycled through local councils or traditional recycling facilities.

  • Reduce the amount of waste that you send to landfill or incinerate
  • Recycle the non recyclable and reduce your carbon footprint
  • Improving your organisation's sustainability can boost your corporate image and improve employee engagement and satisfaction

Use this Zero Waste Box™ to recycle: Safety glasses, face shields and visors.
Do not send in: Other protective equipment
Medical waste. This includes material contaminated with blood, bodily fluids and waste which should be collected in a hazardous clinical waste bin
Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) which presents a danger to the environment or to people
Batteries, pressurised canisters, broken glass

How it works:
• Order - Select a Zero Waste Box™ based on what and how much you would like to recycle.
• Collect - Fill your box with the accepted waste and seal the box with adhesive tape.
• Ship - Using the pre-paid UPS shipping label affixed to the back of your box, send your filled box to TerraCycle® for recycling. UPS requires a minimum of 24 hours’ notice and does pick ups Monday to Friday between 9 am and 5 pm. Pick-ups can be arranged online or by telephone. All shipments are carbon-neutral, meaning the carbon emissions released during transport will be offset.
• Reorder - Continue to recycle and reorder a new Zero Waste Box™. Your collected waste will find a second life as recycled materials, reducing the need to extract virgin materials and depleting resources.

Everything collected through the Zero Waste Box™ system is sorted and processed into raw materials that can be reused instead of being sent to landfill.

Cardboard boxes are made from 84% recycled material. Box container and liner are 100% recyclable.

Caution: Do not send in any item that has come in contact with substances classified as hazardous waste e.g., sharp, flammable, reactive, acidic, poisonous, corrosive, ignitable, toxic, infectious or pathogenic or material contaminated with blood which presents a danger to the environment, or to people.

Do not send in medical waste. This includes material contaminated with blood or bodily fluids and waste that originates from health care facilities, hospitals, home care, physicians' offices, dental offices, blood banks, veterinary clinics, and more.

Specifications
  • Capacity:
    28 L
  • Description:
    Disposal box for protective eyewear, small
  • Seq:
    01
  • W×D×H:
    250×250×460 mm
  • Load capacity:
    ≤7 kg
  • Cat. No.:
    129-2443
  • Environmental certification:
    FSC (Certificate code FSC: BV-COC-150648), Bureau Veritas
  • Environmentally Preferable:
    Waste Reducing|Sustainable Materials