Order Entry
France
ContactUsLinkComponent
Hendra virus Fusion glycoprotein (A263T), His Tag (FUN-H52H4) is expressed from human 293 cells (HEK293). It contains AA Ile 27 - Se r 487 (Accession # O89342-1 (A263T)). 1 * 500 µG
Catalog # ACRBFUN-H52H4500UG
Supplier:  ACROBIOSYSTEMS
Hendra virus Fusion glycoprotein (A263T), His Tag (FUN-H52H4) is expressed from human 293 cells (HEK293). It contains AA Ile 27 - Se r 487 (Accession # O89342-1 (A263T)). 1 * 500 µG
Catalog # ACRBFUN-H52H4500UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together