Order Entry
France
ContactUsLinkComponent
Anti-SARS-CoV-2 Spike RBD Neutralizing Antibody, Chimeric mAb, Cynomolgus IgG1 (AM122) (SPD-M201) is a chimeric monoclonal antibody recombinantly expressed from HEK293 cells, which combines the variable region of a mouse monoclonal antibody with Cyno 1 * 100 µG
Catalog # ACRBSPD-M201-100UG
Supplier:  ACROBIOSYSTEMS
Anti-SARS-CoV-2 Spike RBD Neutralizing Antibody, Chimeric mAb, Cynomolgus IgG1 (AM122) (SPD-M201) is a chimeric monoclonal antibody recombinantly expressed from HEK293 cells, which combines the variable region of a mouse monoclonal antibody with Cyno 1 * 100 µG
Catalog # ACRBSPD-M201-100UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together