Order Entry
France
ContactUsLinkComponent
Human MASP3 (450-721), His Tag (MA3-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ile 450 - Val 721 (Accession # P48740-2). 1 * 100 µG
Catalog # ACRBMA3-H52H3100UG
Supplier:  ACROBIOSYSTEMS
Human MASP3 (450-721), His Tag (MA3-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ile 450 - Val 721 (Accession # P48740-2). 1 * 100 µG
Catalog # ACRBMA3-H52H3100UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together